Finntip™ Flex™ Pipette Tips
Actual product may vary
Finntip™ Flex™ Pipette Tips
Finntip™ Flex™ Pipette Tips
Thermo Scientific
Finntip™ Flex™ Pipette Tips
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Have Questions?
Catalog NumberVolume (Metric)Color CodePackagingFit for Purpose
94060317200 μLYellow1920 tipsCertified Quality
Catalog number 94060317
HSN 84799090
Price (SGD)
64.50
Special offer
Ends: 30-Nov-2026
215.00
Save 150.50 (70%)
Each
Volume (Metric):
200 μL
Color Code:
Yellow
Packaging:
1920 tips
Fit for Purpose:
Certified Quality
Price (SGD)
64.50
Special offer
Ends: 30-Nov-2026
215.00
Save 150.50 (70%)
Each
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
  • Excellent for single and multichannel pipetting
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack
Softer Finntip Flex Tip
  • The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL Extended (bottle tip)
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip Flex 200, Non-Filtered, Non-sterile, Refill
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorYellow
ModelFinntip Flex 200
Length (English)2.3 in.
Length (Metric)6cm
FormatRefill
Packaging1920 tips
Color CodeYellow
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip Flex
Fit for PurposeCertified Quality
Unit SizeEach