Glass Rolled Rim Vials
Glass Rolled Rim Vials
Actual product may vary
Glass Rolled Rim Vials
Glass Rolled Rim Vials are ideal for a variety of laboratory applications. The vials are made of soda-lime glass with a tight closing polyethylene snap-on lid, non-sterile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) Outer
9400230
also known as 9400230IVD
25 mL51 mm30 mm
9400260
also known as FS9400260
30 mL76 mm28 mm
Catalog number 9400230
also known as 9400230IVD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
25 mL
Height (Metric):
51 mm
Diameter (Metric) Outer:
30 mm
  • Soda-lime glass
  • PE snap-on lid
  • Non-sterile
Specifications
Closure ColorNatural
DescriptionRolled rim glass vials with snap-on lid
Diameter (English) Outer1.2 in.
ColorClear
Diameter (Metric) Outer30 mm
MaterialSoda Lime Glass
SterilityNon-sterile
Volume (Metric)25 mL
For Use With (Application)A variety of laboratory applications
Height (English)2 in.
Material (Seal)Polyethylene
Neck StyleRolled Rim
Volume (English)0.8 oz.
Closure MaterialPolyethylene
Closure TypeSnap Cap
Height (Metric)51 mm
Quantity250 Pk.
TypeRolled Rim Vial
Unit SizePK