Orion™ All-in-One™ pH Buffer Kits
Actual product may vary
Orion™ All-in-One™ pH Buffer Kits
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
916099All-in-One pH Buffer Kit
FIS13-641-420ROSS All-in-One pH Buffer Kit
FIS13-624-500Standard All-in-One pH Buffer Kit
Catalog number 916099
HSN 90279090
Price (SGD)
-
Description:
All-in-One pH Buffer Kit
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.
  • Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
  • Orion pH buffers are color-coded for easy identification: pH 4 buffer is pink, pH 7 buffer is yellow and pH 10 buffer is blue.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionAll-in-One pH Buffer Kit
For Use With (Application)Accurate calibrations and NIST traceability, portable and field analysis
pH4.01, 7.00, 10.01
Quantity5 x 60 mL
PackagingKit
Traceability to NISTYes
Includes60 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 60 mL bottle of pH electrode storage solution; and 60 mL bottle of pH electrode cleaning solution
Unit SizeEach