Accessories for Orion™ pH Electrodes
Accessories for Orion™ pH Electrodes
Actual product may vary
Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes with these accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
910003
also known as IC910003
12 mm Electrodes, GlassElectrode Storage Bottle
FIS13-641-4212 mm Electrodes, GlassElectrode Storage Bottle
Catalog number 910003
also known as IC910003
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
12 mm Electrodes, Glass
Type:
Electrode Storage Bottle
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
  • Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
  • Designed for glass body pH electrodes
  • Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
Specifications
Description12mm electrode storage bottle
For Use With (Equipment)12 mm Electrodes, Glass
TypeElectrode Storage Bottle
Unit SizeEach