1x1 image pixel for data collection
Damp Proof Cabinet
Damp Proof Cabinet
Actual product may vary
Damp Proof Cabinet
Keeps works in progress in moist workable condition. Cabinet is air-sealed by foam rubber gasket doors. Doors and all exterior walls are double-walled with an internal layer of anodized sheet aluminum.
Have Questions?
Catalog number 9100
Price (SGD)
-
  • Double-walled with an internal layer of anodized sheet aluminum
  • 36 x 18 x 84 in.
  • 4 Heavy Duty aluminum shelves and water retaining bats
  • Constructed of 19-gauge welded steel with 16-gauge supports
  • Diversified Spaces™ is SEFA compliant and MAS Certified Green
  • Doors have gaskets to keep the clay projects moist and workable
  • Limited Lifetime Warranty
  • Locking doors with a 3 point locking mechanism
  • Made in the U.S.A.
Specifications
Certifications/ComplianceSEFA, Mas Certified Green
Depth (Metric)45 cm
FinishPolished Cast Aluminum
For Use With (Application)Science classrooms
Height (Metric)2.1 m
IncludesLocks
WarrantyLimited lifetime
Weight (English)309 lb.
Weight (Metric)140.1 kg
Width (English)36 in.
Width (Metric)91.4 cm
Product TypeStorage Cabinet
ColorBrown
MaterialAnodized Aluminum
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link