Bottle for Shake and Pour Liquid Diet
Bottle for Shake and Pour Liquid Diet
Actual product may vary
Bottle for Shake and Pour Liquid Diet
Bottle for Shake and Pour Liquid Diet
Bottle for Shake and Pour Liquid Diet
Bottle for Shake and Pour Liquid Diet
For use in preparing Shake and Pour Liquid Diet formulations
Have Questions?
Catalog NumberCapacity (Metric)
9090
also known as 909RAKC0
4700 mL
Catalog number 9090
also known as 909RAKC0
Price (SGD)
-
Capacity (Metric):
4700 mL
  • Graduated, reusable plastic bottle with convenient handle, screw-top lid, and wide mouth
  • 5 qt. (4.7L) size
Specifications
Capacity (Metric)4700 mL
Capacity (English)159 oz.
MaterialPlastic
Closure TypeScrew Cap
Unit SizeEach