Stainless Steel Cylindrical Seamless Scoops
Stainless Steel Cylindrical Seamless Scoops
Actual product may vary
Stainless Steel Cylindrical Seamless Scoops
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Seamless Cylindrical Scoops are particularly suitable for pharmaceutical applications.
Have Questions?
Catalog NumberCapacity (Metric)
9050
also known as ZS9050OSCSHA
1000 mL
Catalog number 9050
also known as ZS9050OSCSHA
Price (SGD)
-
Capacity (Metric):
1000 mL
  • Stainless steel seamless cylindrical scoops 
  • Particularly suitable for pharmaceutical applications  
  • All surfaces are non-reacting 
  • Offer zero absorption suiting GMP requirements
  • Scratches within the scoop prevent cross contamination
Specifications
Product TypeScoop
MaterialStainless Steel
AutoclavableAutoclavable
Capacity (Metric)1000 mL
StyleCylindrical
For Use With (Application)Suitable for Pharmaceutical Applications
Length (Metric)340 mm
Length (Metric) Handle120 mm
Unit SizeEach