Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Actual product may vary
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredFor Use With (Equipment)Fit for Purpose
904-252
also known as 904-252GP
1000 μLNon-filteredCavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
902-25150 μLNon-filteredCavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusSterile
902-25350 μLNon-filteredTecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
904-011
also known as 904-011P
1000 μLFilteredTecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
904-011G1000 μLFilteredTecan Genesis, Tecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
904-253
also known as 904-253GPA
1000 μLNon-filteredTecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer JanusCertified Quality
901-01110 μLFilteredTecan Genesis, Tecan Freedom EVO, MiniPrep, CavroSterile and PCR Ready
Catalog number 904-252
also known as 904-252GP
Price (SGD)
260.00
CS
Add to cart
Volume (Metric):
1000 μL
Filtered:
Non-filtered
For Use With (Equipment):
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for Purpose:
Certified Quality
Price (SGD)
260.00
CS
Add to cart
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
Features:
  • Available as sterile, filtered and/or liquid sensing.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlack
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
FormatRack
Tip StyleConductive
PackagingReload Insert, 96 tips/insert, 10 inserts/pack
Quantity960 Tips
For Use With (Equipment)Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for PurposeCertified Quality
Unit SizeCS