PharmacoScan™ Training Kit, 24-Format
PharmacoScan™ Training Kit, 24-Format
Actual product may vary
PharmacoScan™ Training Kit, 24-Format
Have Questions?
Catalog NumberQuantity
903-01188 samples
Catalog number 903-011
Price (SGD)
352.00
CS
Add to cart
Quantity:
88 samples
Price (SGD)
352.00
CS
Add to cart
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeTraining Kit
ArrayGenotyping
Number of Arrays24 arrays
Format24-array Plate
SpeciesHuman
For Use With (Application)Microarray Analysis
No. of Samples88 samples
Quantity88 samples
Unit SizeCS
Contents & Storage
4 X 24-format array plates. Store at 2-8°C
2 x training plates containing known DNA samples. Store frozen -5 to -30°C.
1 x Reagent Kit (Includes reagents and controls for 4x24-format array plates).Storage can vary for each reagent. Refer to product insert.
Gene Titan instrument consumables for 4x24-format array plates. Store at room temp.