Double Tier Tube Platform
Double Tier Tube Platform
Actual product may vary
Double Tier Tube Platform
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Have Questions?
Catalog number 88882158
Price (SGD)
133.00
EA
Add to cart
Price (SGD)
133.00
EA
Add to cart
Thermo Scientific TM Double Tier Tube Platform designed for use with the Digital Cel-Gro Tissue Culture Rotator accommodate double capacity in a compact footprint.
Specifications
Tube Size30 mm
MaterialMetal
Dimensions (L x W)33 x 14 x 1.2 cm (13 x 5.5 x 0.5 in.)
Dimensions (L x W x H)33 x 14 x 1.2 cm (13 x 5.5 x 0.5 in.)
DescriptionDouble Tier Tube Platform
For Use With (Application)diameter 5-30 mm tubes
For Use With (Equipment)Digital Tube Rocker, 120V (88882017), Digital Tube Rocker, 230V (88882018)
No. of Positions7
Unit SizeEA