1x1 image pixel for data collection
Micropump™ Gear Pump Head 185
Micropump™ Gear Pump Head 185
Actual product may vary
Micropump™ Gear Pump Head 185
Smooth pulseless flow provides precise, leak-free delivery of fluids.
Have Questions?
Catalog NumberFlow Rate
888821155 to 504 mL/min
Catalog number 88882115
Price (SGD)
32.00
EA
Estimated availability date 16-Nov-2025
Add to cart
Flow Rate:
5 to 504 mL/min
Price (SGD)
32.00
EA
Add to cart
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
  • Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery.
  • The magnetic drive and static seals keep fluids securely inside and potential contaminants out.
  • Chemical resistant materials handle a wide range of materials (due to wetted parts).
  • Pump heads are compact and easy to service.
  • The modular design offers flexibility—configure to your specific needs.
Specifications
Flow Rate5 to 504 mL/min
Product Type96-well Microplate Clamps
MaterialStainless Steel
Dimensions (L x W x H)54 x 19 x 19 mm
Description96-well microplate clamps
For Use With (Equipment)Rocker, Rotator, mixer
Includes7 Clamps
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link