Basic Vortex Mixers
Basic Vortex Mixers
Actual product may vary
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
Basic Vortex Mixers
The Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.
Have Questions?
Catalog NumberCertifications/CompliancePlug Type
88882012CEEU, UK, CN Plug
Catalog number 88882012
Price (SGD)
488.00
EA
Add to cart
Certifications/Compliance:
CE
Plug Type:
EU, UK, CN Plug
Price (SGD)
488.00
EA
Add to cart
The Thermo Scientific™ Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.
  • Digital Speed display 
  • Continuous and Touch modes
  • Speed range: 200 to 3000rpm
  • Speed Accuracy: ± 5% rpm
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels
Specifications
Orbit4 mm
Load Bearing Capacity (Metric)0.5 kg
Speed Range200 to 3000 rpm
Electrical Requirements100/240 V
HoldsTubes
Certifications/ComplianceCE
Plug TypeEU, UK, CN Plug
Frequency50/60 Hz
MaterialABS housing
Dimensions (L x W x H)290 x 270 x 240 mm (11.4 x 10.7 x 9.4 in.)
Length (English)11.4 in.
Length (Metric)290 mm
Shipping Weight (English)14.99 lb.
Shipping Weight (Metric)6.8 kg
Weight (English)14.33 lb.
Weight (Metric)6.5 kg
ModelBasic Vortex Mixer
DisplayDigital
For Use With (Equipment)Accessories: 88882120, 88882121, 88882122, 88882123, 88882124, 88882125, 88882126, 88882127
IncludesRubber platform 88882120
IP RatingIP 32
Motor TypeDC motor
No. of Positions19
Operating ModesTouch, Continous
Warranty and Service Offering2 Years
Unit SizeEA