1x1 image pixel for data collection
Digital Mini Rotators
Digital Mini Rotators
Actual product may vary
Digital Mini Rotators
Digital Mini Rotators
Digital Mini Rotators
Digital Mini Rotators
Digital Mini Rotators
Digital Mini Rotators
Digital Mini Rotators
Get accurate and efficient sample mixing
Have Questions?
Catalog NumberPlug TypeHeight (English)Voltage
88882008EU, UK, CN Plug5.1 in.230 V
Catalog number 88882008
Price (SGD)
1,400.00
Special offer
1,700.00
Save 300.00 (18%)
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Height (English):
5.1 in.
Voltage:
230 V
Price (SGD)
1,400.00
Special offer
1,700.00
Save 300.00 (18%)
EA
Add to cart
Provides smooth, accurate and orbital mixing of various vessels in the lab
  • Speed range: 50 – 500 rpm
  • Precise speed control:
  • ±1 rpm (≤100rpm)
  • ±1% (101 - 450rpm)
  • Orbit Diameter: 10mm
  • Max Load up to 4.5 kg
  • 2 Timer modes: Continuous, Timer
  • Multi-platform choices for loading all sizes vessels in the labs.
Specifications
Orbit10 mm
Load Bearing Capacity (Metric)4.5 kg
Speed Control Accuracy±1 rpm (≤100 rpm), ±1% (101 to 500 rpm)
Speed Range50 to 500 rpm
Voltage230 V
Amperage40 VA
Product TypeDigital Mini Rotator
Load Bearing Capacity (English)9.92 lb.
Certifications/CompliancecCSAus, CE, RoHS
Plug TypeEU, UK, CN Plug
Frequency50/60 Hz
Dimensions (D x W x H) Interior383 x 284 x 130 mm (15.1 x 11.2 x 5.1 in.)
Dimensions (LxW) Platform310 x 284 mm (12.2 x 11.2 in.)
Dimensions (L x W x H)487 x 387 x 264 mm (19.2 x 15.2 x 10.4 in.)
Height (English)5.1 in.
Height (Metric)130 mm
Shipping Weight (English)36.2 lb.
Shipping Weight (Metric)16.4 kg
Controller TypeLED
ModelDigital Mini Rotator
DisplayLED
Humidity Range20 to 85%
Timer Range1 min. to 99:59 hr., Continuous
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link