Digital Waving Rotators
Digital Waving Rotators
Actual product may vary
Digital Waving Rotators
Digital Waving Rotators
Digital Waving Rotators
Digital Waving Rotators
Digital Waving Rotators
Get accurate and efficient sample mixing
Have Questions?
Catalog NumberPlug TypeVoltage
88882004EU, UK, CN Plug230 V
Catalog number 88882004
Price (SGD)
2,127.00
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Voltage:
230 V
Price (SGD)
2,127.00
EA
Add to cart
360° orbital waving rotator for efficient sample mixing with accurate speed control and easy angle adjustment
  • Digital display of Speed and Time
  • Speed range: 5 – 120 rpm
  • Easily adjustable rocking angle: 1°- 12°
  • Precise speed control:
  • ±1 rpm ≤ 30 rpm
  • ±2 rpm > 30 rpm
  • Max load up to 9.8 kg
  • Tray size fits 6 x well-plates
  • Timer Range: 1min to 99hrs 59min
  • Optional Non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags
Specifications
Load Bearing Capacity (Metric)9.8 kg
Speed Control Accuracy1 rpm
Speed Range5 to 120 rpm
Electrical Requirements100/240 V
Voltage230 V
Temperature Range (Metric)5°C to 40°C
Amperage0.2 A
Product TypeDigital Waving Rotator
Load Bearing Capacity (English)22 lb.
Certifications/CompliancecCSAus, CE, RoHS
Plug TypeEU, UK, CN Plug
Temperature Range (English)41°F to 104°F
Frequency50/60 Hz
MaterialStainless Steel Platform
Dimensions (D x W x H) Interior383 x 295 x 224 mm (15.1 x 11.6 x 8.8 in.)
Dimensions (LxW) Platform316 x 276 mm (12.4 x 10.9 in.)
Dimensions (L x W x H)487 x 387 x 354 mm (19.2 x 15.2 x 13.9 in.)
Length (English) Platform12.4 in.
Length (Metric) Platform316 mm
Width (English) Platform10.9 in.
Width (Metric) Platform276 mm
Shipping Weight (English)34.8 lb.
Shipping Weight (Metric)15.8 kg
Weight (English)29.8 lb.
Weight (Metric)13.5 kg
Controller TypeLED
ModelDigital Waving Rotator
DisplayLED
For Use With (Equipment)Rocker, Rotator, mixer
Humidity Range20 to 80%
Platform StyleTray
Timer Range1 min. to 99:59 hr.
Unit SizeEA