Racks and Accessories for CO2 Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDimensions (L x W)
88881126Sticky Mat140 x 140 mm
88881119Fixed Tube Rack40 x 16 mm dia.
88881121Adjustable Tube Rack40 x 16 mm dia.
88881122Universal Platform360 x 300 mm
88881123Rubber PlatformN/A
8888112496-well Microplate HolderN/A
88881125Spring Wire Rack240 x 350 mm
88881127Control Box158 x 95 mm
88881128General Power AdapterN/A
88881132UK PlugN/A
Catalog number 88881126
HSN 84798200
Price (SGD)
74.00
Special offer
Ends: 31-Mar-2026
76.00
Save 2.00 (3%)
Each
Add to cart
Product Type:
Sticky Mat
Dimensions (L x W):
140 x 140 mm
Price (SGD)
74.00
Special offer
Ends: 31-Mar-2026
76.00
Save 2.00 (3%)
Each
Add to cart
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Product TypeSticky Mat
Dimensions (L x W)140 x 140 mm
For Use With (Equipment)CO2 Resistant Shaker
No. of Positions4
Unit SizeEach