Accessories for Tube Revolver
Accessories for Tube Revolver
Actual product may vary
Accessories for Tube Revolver
Optimize the performance of your Thermo Scientific™ Tube Revolver by using these accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
88881009Paddles 50mL
88881007Paddles 1.5 mL/2mL
88881005Paddles 10mL/15 mL
88881008Paddles 0.5mL/0.8mL
Catalog number 88881009
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Paddles 50mL
Optimize performance by using Thermo Scientific™ Accessories for Tube Revolver.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
HoldsTubes
Tube Capacity50 mL
Product TypeRotisserie Paddle
DescriptionPaddles 50mL
For Use With (Equipment)Rotator/Rocker
Unit SizeEA