Tube Revolver Rotator
Tube Revolver Rotator
Actual product may vary
Tube Revolver Rotator
Increase your lab's efficiency with our variable speed tube rotator with a small footprint, flexible design, and simple display that help increase flexiblity and effectiveness.
Have Questions?
Catalog NumberHoldsSpeed RangePlug Type
88881002Combination of 4 paddles of:
7 x 10 to 15 mL tubes
7 x 5 to 7 mL tubes
15 x 1.5 to 2 mL tubes
21 x 0.5 to 0.8 mL tubes
One paddle of 6 x 50 mL tubes
10 to 40 rpmCord set with various plugs
Catalog number 88881002
Price (SGD)
654.00
EA
Add to cart
Holds:
Combination of 4 paddles of:
7 x 10 to 15 mL tubes
7 x 5 to 7 mL tubes
15 x 1.5 to 2 mL tubes
21 x 0.5 to 0.8 mL tubes
One paddle of 6 x 50 mL tubes
Speed Range:
10 to 40 rpm
Plug Type:
Cord set with various plugs
Price (SGD)
654.00
EA
Add to cart
Increase your lab's efficiency with the variable speed Thermo Scientific™ Tube Revolver / Rotator. A small footprint, flexible design and simple display help increase flexiblity and effectiveness.
  • Interchangeable rotisseries accommodate different tube sizes for maximum flexiblity. Each paddle can be adjusted from 0-90 degrees.
  • Achieve more effective mixing with oscillating mode. Oscillation will occur at every 40 degrees in the rotation cycle.
  • Small footprint
  • Plug-and-play simplicity
  • Maintenance-free motor, quiet operation
Warranty: 2 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed Range10 to 40 rpm
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Temperature Range (Metric)4°C to 60°C
Wattage6 W
HoldsCombination of 4 paddles of:
7 x 10 to 15 mL tubes
7 x 5 to 7 mL tubes
15 x 1.5 to 2 mL tubes
21 x 0.5 to 0.8 mL tubes
One paddle of 6 x 50 mL tubes
Product TypeRotator
Certifications/CompliancecCSAus, CE
Plug TypeCord set with various plugs
Frequency50/60 Hz
Dimensions (L x W x H)7.7 in., 195 mm
Height (English)7.7 in.
Height (Metric)195 mm
Length (English)6 in.
Length (Metric)154 mm
Width (English)8.9 in.
Width (Metric)225 mm
Shipping Weight (English)2.65 lb.
Shipping Weight (Metric)1.2 kg
ModelTube Revolver, Rotator
IncludesTwo 10/15mL paddles (88881005)
Two 5mL/7mL paddles (88881006)
Two 1.5mL/2mL paddles (88881007)
One 50mL paddle (88881009)
Paddles can also be purchased as accessories in quantities listed above
Warranty and Service Offering2 Years
Unit SizeEA