Block, 3 x 50 mL
Product Image
Block, 3 x 50 mL
For Use With: Thermo Scientific™ Compact Dry Bath/Block Heater
Have Questions?
Catalog NumberNo. of Places
888711063
Catalog number 88871106
Price (SGD)
177.00
EA
Add to cart
No. of Places:
3
Price (SGD)
177.00
EA
Add to cart
Increase your application capabilities with a variety of accessories for the Thermo Scientific™ Compact Dry Baths/Block Heaters.
Warranty: 2 years
Specifications
Description3 x 50mL block
Capacity (Metric)50 mL
For Use With (Equipment)Thermo Scientific™ Compact Dry Bath/Block Heater
LightingLED
No. of Places3
TypeDry Block
Dimensions (L x W x H)36 x 108 x 72 mm (1.4 x 4.3 x 2.8 in.)
Unit SizeEA