Pharma scoop
Product Image
Pharma scoop
Bochem™ Pharma scoop is a highly polished scoop with a handle.
Have Questions?
Catalog NumberCapacity (Metric)
8860100 mL
Catalog number 8860
Price (SGD)
811.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
811.00
EA
Add to cart
  • Polished
  • Includes handle
Specifications
MaterialStainless Steel (316)
Description1.22mm wall thickness
Capacity (Metric)100 mL
Length (Metric)180 mm
Length (Metric) Bowl100 mm
Width (Metric) Bowl50 mm
Unit SizeEA