CryoBoxes™
CryoBoxes™
Actual product may vary
CryoBoxes™
CryoBoxes™
CryoBoxes™
CryoBoxes™
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsMaterialUnit Size
867013-0245
also known as NLG867013-0245
Assorted81 (1 to 2 mL Vials)PolycarbonatePK
330821White64 (1.5 mL Microcentrifuge Tubes)PolycarbonateCS
FIS12-565-227White64 (1.5 mL Microcentrifuge Tubes)PolycarbonatePK
FIS12-567-04White100 (3.6 mL Vials)CardboardCS
341483
also known as NNU341483
White100 (3.6 mL Vials)PolycarbonatePK
FIS12-565-234White100 (3.6 mL Vials)PolycarbonatePK
NNU341483White100 (3.6 mL Vials)PolycarbonatePack
369639White81 (1 to 1.8 mL Vials)CardboardCS
FIS12-567-02White81 (3.6 mL Vials)CardboardCS
374187Gray100 (1 to 1.8 mL Vials)PolycarbonateCS
FIS12-565-182Brown81 (1 to 1.8 mL Vials)CardboardCS
5025-0505
also known as NLG5025-0505
White25 (1.2 and 2 mL Vials)PolycarbonateCS
FIS15-350-107White25 (1.2 and 2 mL Vials)PolycarbonatePK
FKS10739331White25 (1.2 and 2 mL Vials)PolycarbonatePK
5026-0909White81 (1.2 and 2 mL Vials)PolycarbonateCS
FIS15-350-107BWhite81 (1.2 and 2 mL Vials)PolycarbonatePK
5026-1010
also known as 5026-1010P
Blue100 (1 to 1.5 mL Vials)PolycarbonateCS
FIS03-337-7AABlue100 (1 to 1.5 mL Vials)PolycarbonateCS
FIS03-337-7JWhite81 (5 mL Vials)PolycarbonatePK
867013-0242
also known as NLG867013-0242
Green81 (1 to 2 mL Vials)PolycarbonatePK
867013-0243
also known as NLG867013-0243
Blue81 (1 to 2 mL Vials)PolycarbonateCS
FIS14-175-65Assorted81 (1 to 2 mL Vials)PolycarbonateCS
FKS10151042Assorted81 (1 to 2 mL Vials)PolycarbonatePK
Catalog number 867013-0245
also known as NLG867013-0245
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Assorted
Holds:
81 (1 to 2 mL Vials)
Material:
Polycarbonate
Unit Size:
PK
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimized to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
  • Durable, all-plastic CryoBoxes™ for short- or long-term storage
  • Keyed lids on plastic CryoBoxes™ prevent misalignment
  • Plastic boxes are autoclavable for decontamination and usable at temperature range from -196°C to +121°C
  • Printed grid on plastic lids for easy inventory management
  • Economical chipboard/cardboard CryoBoxes™ for short term storage
  • Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
  • Available with dividers from 25 to 169 tube storage
Specifications
DescriptionPolycarbonate, 133 x 133 x 52mm
ColorAssorted
MaterialPolycarbonate
ShapeSquare
TypeCryo Box
Holds81 (1 to 2 mL Vials)
IncludesColored Insert
Product LineNalgene
Quantity4/Pk., 24/Cs.
Tube Array9 x 9
Dimensions (L x W x H)5.25 x 5.25 x 2 in.
Length (English)5.25 in.
Length (Metric)133 mm
Width (English)5.25 in.
Width (Metric)133 mm
Height (English)2 in.
Height (Metric)52 mm
Weight (Metric)4.3 kg
Unit SizePK