FlipStrip™ Microtube Racks with Lid
FlipStrip™ Microtube Racks with Lid
Actual product may vary
FlipStrip™ Microtube Racks with Lid
Hold 0.2mL strips and 1.5mL tubes on one side and and 0.5mL tubes on the other side of this microtube rack with lid, which is made from 100% recycled plastic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
8668Amber, Amber
FKS12368182Yellow
Catalog number 8668
Price (SGD)
37.00
PK
Add to cart
Color:
Amber, Amber
Price (SGD)
37.00
PK
Add to cart
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
  • Several colors available
Specifications
Certifications/ComplianceZZ-T-831D Types I and III, U.S. Pharmacopeia Class VI
Product TypeFlipstrip Rack, Flipstrip Rack
AutoclavableAutoclavable
Diameter (English) Inner0.156 in.
Diameter (Metric) Inner4 mm
MaterialRubber, Rubber
ColorAmber, Amber
Diameter (English) Outer1/4 in.
Diameter (Metric) Outer6.4 mm
HoldsMicrotubes (0.5 mL), Microtubes (1.5 mL), Microtubes (2 mL)
Overall Length (English)12 ft.
Overall Length (Metric)3.7 m
Thickness (English)0.047 in.
Thickness (Metric)1.2 mm
ShapeRectangular
StackableYes
No. per Case12 Racks
Unit SizePK