18/10 Stainless Steel High Shape Laboratory Bowl
18/10 Stainless Steel High Shape Laboratory Bowl
Actual product may vary
18/10 Stainless Steel High Shape Laboratory Bowl
Manufactured from 18/10 stainless steel
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
860110 L150 mm80 mm
Catalog number 8601
Price (SGD)
20.00
EA
Add to cart
Capacity (Metric):
10 L
Diameter (Metric):
150 mm
Height (Metric):
80 mm
Price (SGD)
20.00
EA
Add to cart
  • High shape
  • With rim
  • 18/10 stainless steel
Specifications
Bottom ShapeFlat
Capacity (Metric)10 L
MaterialStainless Steel
ShapeRound
TypeLaboratory Bowl
Height (Metric)80 mm
Diameter (Metric)150 mm
Unit SizeEA