TSKgel™ Super-Octyl HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™
Actual product may vary
Thermo Scientific
TSKgel™ Super-Octyl HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ Super-Octyl Columns are packed with monodispersed, 2 μm (nominal, mean particle size is 2.3 μm) spherical silica particles covalently bonded with octyl groups.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
8200135 cm2 mm
Catalog number 820013
HSN 39231090
Price (SGD)
52.00
Each
Length (Metric):
5 cm
Diameter (Metric):
2 mm
Price (SGD)
52.00
Each
  • A small particle size makes the Super series the highest efficiency, reversed phase columns in the TSKgel product line
  • Monodispersed packing generates operational back pressures more typical of larger particles, allowing the use of higher flow rates than other 2 μm packings
  • Offers less hydrophobicity than TSKgel Super-ODS
  • An excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules, and food/beverage samples
TSKgel is a registered trademark of Tosoh Corporation
Specifications
Diameter (Metric)2 mm
Flow Rate0.25 mL/min
Length (Metric)5 cm
Max. Pressure150 bar
Particle ShapeSphere
Particle Size2.3 μm
pH2 to 7.5
Pore Size110 Å
Solvents0-100% Organic Solvent
Surface Area100 m2/g
Temperature10°C to 50°C
USP TypeL7
Stationary PhaseC8 (octyl)
Product LineTSKgel
Shipping ConditionShipped in 70% Methanol, 30% Water
MatrixC8 (octyl) Phase/Silica Particle Platform
Carbon Load5%
EndcappedYes
PhaseReversed Phase
MaterialStainless Steel
TechniqueReversed Phase HPLC
Unit SizeEach