Orion™ pH Electrode Filling Solution
Orion™ pH Electrode Filling Solution
Actual product may vary
Orion™ pH Electrode Filling Solution
Use this pH electrode filling solution with pH electrodes and reference electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFilling Solution TypeQuantity
810007Surfactant Electrode Fill5 x 60 mL
FIS13-301-131pH Electrode Fill1 x 60 mL
FIS13-641-755Surfactant Electrode Fill5 x 60 mL
FIS13-641-901Reference Inner Fill5 x 60 mL
FIS13-641-686AgCl Fill Solution5 x 60 mL
FIS13-641-845pH Electrode Fill5/Cs.
FKS13499216KCl Fill Solution5 x 60 mL
Catalog number 810007
Price (SGD)
-
Filling Solution Type:
Surfactant Electrode Fill
Quantity:
5 x 60 mL
Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.
  • Only use ROSS™ filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide
Warranty: Out of box.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFilling Solution for ROSS Ultra, ROSS, PerpHecT ROSS Electrodes
For Use With (Equipment)ROSS Ultra, ROSS, PerpHecT ROSS Electrodes
TypepH Electrode Filling Solution
Filling Solution TypeSurfactant Electrode Fill
PackagingBottle
Quantity5 x 60 mL
Unit SizeEach