HEPA Filter
HEPA Filter
Actual product may vary
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
HEPA Filter
For Use With: For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series Incubators
Have Questions?
Catalog number 760209
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Specifications
DescriptionMain HEPA Filter, Value pack (4/pk)
For Use With (Equipment)Forma™ Steri-Cycle™, Direct Heat and Water Jacket
LightingLED
TypeHEPA Filter
Unit SizeKT