6-Place Fixed-Angle Highconic™ Rotor Adapters
6-Place Fixed-Angle Highconic™ Rotor Adapters
Actual product may vary
6-Place Fixed-Angle Highconic™ Rotor Adapters
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube Type
76002905
also known as 76002905FS
Microtube
75003102Conical
75003103Conical Falcon
Catalog number 76002905
also known as 76002905FS
Price (SGD)
219.00
EA
Add to cart
Tube Type:
Microtube
Price (SGD)
219.00
EA
Add to cart
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
  • The adapters accommodate round- or conical-bottom tubes and microtubes.
  • Rotor adaptors work with HiGhConic™ fixed angle rotors
  • Fits tube sizes up to 50mL
Specifications
Tube TypeMicrotube
For Use With (Equipment)HiGhConic™ fixed angle rotor
Tube Size1.5 mL
Unit SizeEA