General Purpose Centrifuge Rotor Packages
Actual product may vary
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
75016108TX-400 Cell Culture Package
75016105TX-1000 Cell Culture Package
75016107TX-400 Blood Tube Package
75016110TX-750 Cell Culture Package
Catalog number 75016108
Price (SGD)
4,039.00
Special offer
Ends: 30-Jun-2026
4,751.00
Save 712.00 (15%)
Each
Model:
TX-400 Cell Culture Package
Price (SGD)
4,039.00
Special offer
Ends: 30-Jun-2026
4,751.00
Save 712.00 (15%)
Each
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
MaterialSee individual centrifuge, rotor and accessory data
TypeRotor Equipment Package
CapacityMax in TX-400: 36 x 15 mL Conical Tubes, 16 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net WeightSee individual components of package
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/ComplianceISO 14971, ISO 13485, ISO 9001
IncludesTX-400 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters
WarrantySee Manual
BiocontainmentYes
ModelTX-400 Cell Culture Package
Unit SizeEach