Check Order Status
Medifuge Benchtop Centrifuge Spacers
Actual product may vary
Medifuge Benchtop Centrifuge Spacers
Get accessories for Medifuge small centrifuge
Have Questions?
Catalog Number
Length (Metric)
Color
75008817
2 cm
Green
Catalog number
75008817
Price (SGD)
97.00
EA
Add to cart
Length (Metric):
2 cm
Color:
Green
Price (SGD)
97.00
EA
Add to cart
Get accessories for Medifuge small centrifuge
Specifications
Type
Benchtop Centrifuge Spacer
For Use With (Equipment)
Medifuge Benchtop Centrifuge
Color
Green
Capacity
13 mL
Description
Spacer
Length (Metric)
2 cm
Includes
Set of 8