Check Order Status
Round Buckets for 4-Place Swinging Bucket Rotor
Round Buckets for 4-Place Swinging Bucket Rotor
4 x 100mL capacity buckets fit the Thermo Scientific Swinging 4-place rotor (75-007-591)
Have Questions?
Catalog Number
For Use With (Equipment)
75007555
Swinging bucket rotor
Catalog number
75007555
Price (SGD)
686.00
EA
Add to cart
For Use With (Equipment):
Swinging bucket rotor
Price (SGD)
686.00
EA
Add to cart
Specifications
For Use With (Equipment)
Swinging bucket rotor