Sorvall LYNX 4000 Superspeed Centrifuge
Sorvall LYNX 4000 Superspeed Centrifuge
Actual product may vary
Sorvall LYNX 4000 Superspeed Centrifuge
Perform high-speed separations simply in shared lab settings with the Sorvall LYNX 4000 superspeed centrifuge with 4L capacity and performance up to 68,905 x g.
Have Questions?
Catalog NumberElectrical RequirementsNet WeightDescription
75006580200/240 V 50/60 Hz 30 A, Single Phase622 lb. (283 kg)Sorvall LYNX 4000 Superspeed Centrifuge
Catalog number 75006580
Price (SGD)
57,854.00
EA
Add to cart
Electrical Requirements:
200/240 V 50/60 Hz 30 A, Single Phase
Net Weight:
622 lb. (283 kg)
Description:
Sorvall LYNX 4000 Superspeed Centrifuge
Price (SGD)
57,854.00
EA
Add to cart
Perform high-speed separations simply in shared lab settings with the Thermo Scientific™ Sorvall™ LYNX 4000 superspeed centrifuge with 4L capacity and performance up to 68,905 x g. Advanced rotor innovations including Thermo Scientific™ Auto-Lock™ rotor exchange, Thermo Scientific™ Auto-ID™ instant rotor identification and lightweight and durable Thermo Scientific™ Fiberlite™ carbon fiber rotors that shorten run set-up time and increase rotor security. For real-time run monitoring and control via a smart device, download the Thermo Scientific™ Centri-Vue™ application.
From academic research to bioproduction, support your high performance separations and evolving application needs:
  • Exceptional performance up to 68,905 x g and up to 4L capacity
  • Shortened run set-up time and increased rotor security with rotor innovations
    • Quickly and securely change rotors and applications in as little as 3 seconds with push-button Auto-Lock rotor exchange
    • Auto-ID instant rotor identification automatically loads rotor specifications into centrifuge parameters
  • Choose from 16 optional rotors to find the ideal fit for your application, supporting labware from microplates, to microtubes, to 1L bottles:
    • Lightweight Fiberlite carbon fiber rotors with corrosion- and fatigue-resistance secure the rotor's structural integrity, backed by a 15-year warranty
    • Swinging bucket rotors for versatility and expanded capacity at high RCF with dozens of adapters for research flexibility
    • Complete fixed angle rotor portfolio for commonly used 50mL, 50mL conical, 250mL, 500mL and 1L tubes and bottles with specialty rotor options to address specific protocol requirements
  • Centri-Touch touchscreen interface with bright, highly visible, and durable display simplifies run set-up while providing on-board training and access controls, such as user login with password protection
  • Download the Centri-Vue application for real-time run monitoring and control of 1 or up to 100 centrifuges from your mobile device
  • Advanced technology for resource efficiency including energy use options
  • Compact, non-bolt down design, automatic door opening, and an integrated surface work space optimize everyday centrifuge use
  • Compliant with global standards and designed to assist with GMP/GLP data management
    • Compatible with Centri-Log Plus data management software for FDA 21 CFR Part 11 compliance
Specifications
Depth (English) Exterior31.7 in.
Depth (Metric) Exterior805 mm
Frequency50/60 Hz
Profile (Acceleration/Braking)9 Accel, 10 Decel
TypeSuperspeed Centrifuge
Capacity4 x 1000 mL
Max. Speed24,000 rpm
Max. RCF68,905 x g
RefrigeratedYes
Height (English) Exterior36.6 in.
Height (Metric) Exterior930 mm
Length (English) Exterior31.7 in.
Length (Metric) Exterior805 mm
Noise Level<59 dBA
Width (English) Exterior27.6 in.
Width (Metric) Exterior700 mm
DescriptionSorvall LYNX 4000 Superspeed Centrifuge
Drive SystemHigh Torque Brushless
Green TechnologyGreen Mode
Other FunctionsMultilingual Selection, Rotor Calculator, On-Board Training Videos, User Logging, User Lock-Out, Automatic Door Opening, Rotor Speed Handle
Run Time99 hours, hold
WarrantyUnit: 1 yr., Drive: 3 yrs., Refrigeration: 5 yrs
Electrical Requirements200/240 V 50/60 Hz 30 A, Single Phase
Certifications/CompliancecULus, CE, ROHS, WEEE
Controller TypeTouchscreen Interface
Dimensions (HWD)36.6 x 27.6 x 31.7 in. (93.0 x 70.0 x 80.5 cm)
Net Weight622 lb. (283 kg)
Program Storage120 programs with alpha-numeric naming
StandardsIEC 61010-2-020 2nd Ed. or later
Temperature Range-10°C to +40°C
Product LineSorvall
Rotor IdentificationAuto-ID Instant
Green FeaturesEnergy efficient, less waste
Unit SizeEA
Frequently asked questions (FAQs)
What rotor does the Sorvall LYNX 4000 Superspeed Centrifuge (Cat. No. 75006580) come with?
The Sorvall LYNX 4000 Superspeed Centrifuge (Cat. No. 75006580) does not come with any specific rotors included. However, it is compatible with a variety of rotors to fit different applications, supporting labware from microplates, to microtubes, to 1 L bottles (https://assets.thermofisher.com/TFS-Assets/LED/Specification-Sheets/Superspeed-Centrifuge-Rotors-RTBA-EN.pdf).