Check Order Status
Adapters for HIGHConic III Rotor
Actual product may vary
Adapters for HIGHConic III Rotor
Assorted adapters to be used with the HIGHConic III rotor
Have Questions?
Change view
Catalog Number
Capacity
Max. Tube Diameter
Max. Tube Length
75005808
8 x 15 mL
12 mm
17 mm
75005807
1.5/2.0 mL
11 mm
40 mm
75005770
5 mL
17 mm
100 mm
3 Options
Catalog number
75005808
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
8 x 15 mL
Max. Tube Diameter:
12 mm
Max. Tube Length:
17 mm
Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.
Adapters to run various tubes in HIGHConic III rotor
Specifications
Type
Rotor Adapter
Tube Type
Conical
Capacity
8 x 15 mL
Max. Tube Diameter
12 mm
Max. Tube Length
17 mm
For Use With (Equipment)
HIGHConic III fixed angel rotor (75005709).
Description
15mL Conical Tube Adapter
Tube Size
15 mL