Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Actual product may vary
Thermo Scientific
Buckets for Thermo Scientific™ M10 Microplate Swinging Bucket Rotor
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
75005723Unsealed Bucket
75005721Sealed Bucket
Catalog number 75005723
Price (SGD)
1,336.00
EA
Add to cart
Type:
Unsealed Bucket
Price (SGD)
1,336.00
EA
Add to cart
Accessories for the Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Max Tube Dimensions: Height <33 mm
Specifications
For Use With (Equipment)Thermo Scientific™ M10 Microplate Swinging Bucket Rotor.
Capacity4 Standard or 2 Midi-Deepwell plates
TypeUnsealed Bucket
DescriptionUnsealed Buckets (set of 2)
Unit SizeEA