Sorvall™ ST 40 Centrifuge Series
Sorvall™ ST 40 Centrifuge Series
Actual product may vary
Sorvall™ ST 40 Centrifuge Series
Process 40% more 50mL conical tubes or 100% more blood tubes with biocontainment sealing than competitive models with the Sorvall ST 40 Centrifuge series.
Have Questions?
Catalog NumberRefrigeratedElectrical RequirementsDescription
75004524Yes230 V 50/60 HzSorvall ST 40R Centrifuge
Catalog number 75004524
Price (SGD)
-
Refrigerated:
Yes
Electrical Requirements:
230 V 50/60 Hz
Description:
Sorvall ST 40R Centrifuge
Process 40% more 50mL conical tubes or 100% more blood tubes with biocontainment sealing than competitive models with the Thermo Scientific™ Sorvall™ ST 40 Centrifuge Series, which offers exceptional capacity, ergonomic features and value for routine sample processing on the benchtop. The innovative design enables safe, simple and comfortable operation with marketing-leading capabilities.
Easy to Use:
  • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
  • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
Thermo Scientific Auto-Lock™ III Rotor System:
  • Secure locking system allows easy push-button installation and exchange of rotors.
  • System saves time when switching between applications.
  • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Thermo ScientificClickSeal™ Bucket Sealing System:
  • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
  • System has glove-friendly, one-handed open/close capability.
  • System is certified by CAMR™ in Porton Down, UK.
Thermo ScientificSMARTSpin™ Technology:
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
  • Technology optimizes safety and improves separations.
Space-Saving Design:
  • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
Outstanding Energy Efficiency:
  • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Ventilated Model ST 40:
  • Dimensions: 26.4L × 22.2W × 14.2 in.H [35.4 in.H open] (67 × 56.5 × 36cm [90cm open])
  • Net weight: 189.6 lb. (86kg)
Refrigerated Model ST 40R:
  • CFC-free refrigeration system
  • Temperature Range: -10° to +40°C
  • Pre-Cooling Function with direct button
  • 26.4L × 29.3W × 14.2 in.H [35.4 in.H open] (67 × 74.5 × 36cm [90cm open])
  • Net weight: 255.7 lb. (116kg)
Ordering Information:
A wide range of rotors are available for your daily routine processing requirements. Maximize performance with lighter-weight Thermo Scientific Fiberlite carbon fiber rotors that offer versatility, speed and robust, corrosion-free design. All rotors and adapters are sold separately.
Recommended for:
  • Cell Culture
  • Blood Processing
Specifications
Certifications/ComplianceIVD Compliant, IEC 61010-1, IEC 61010-2-202, IEC 61010-2-101. CSA, Certified Biosafety, CE marked, UL listed.
Controller TypeMicroprocessor
DescriptionSorvall ST 40R Centrifuge
Capacity4 x 1000 mL
Max. RCF25,314 x g
Max. Speed15,200 rpm
Product LineSorvall
RefrigeratedYes
DisplayDigital
Drive SystemDirect, Brushless Induction Low Profile
Noise Level<61 dB(A)
Profile (Acceleration/Braking)9 Accel/10 Braking
Program Storage6 programs, direct access
Run Time9 hr. 99 min. plus HOLD
TypeCentrifuge
Electrical Requirements230 V 50/60 Hz