TX-400 Rotor Adapters
TX-400 Rotor Adapters
Actual product may vary
TX-400 Rotor Adapters
For use with TX-400 Swinging Bucket Rotor
Have Questions?
Catalog NumberFor Use With (Equipment)
75003825TX-400 Swinging Bucket Rotor
Catalog number 75003825
Price (SGD)
1,569.00
EA
Add to cart
For Use With (Equipment):
TX-400 Swinging Bucket Rotor
Price (SGD)
1,569.00
EA
Add to cart
For use with TX-400 Swinging Bucket Rotor
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 x 250mL (4 x 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. x L: 62 x 125mm (80 x 125mm with 400mL bio-bottles)
Specifications
TypeRotor Adapter
Capacity4.5/6 mL
Max. Tube Diameter13 mm
Max. Tube Length110 mm
For Use With (Equipment)TX-400 Swinging Bucket Rotor
Tube Size4.5/6 mL
Unit SizeEA