1x1 image pixel for data collection
TX-200 Rotor Tube Adapters
TX-200 Rotor Tube Adapters
Actual product may vary
TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500381550 mL Round-bottom Tube
7500377115 mL Conical Tube
750037855/7 mL Blood Collection Tube
7500380350 mL Conical Tube
7500380810 mL Blood Collection or 15 mL Corex /Kimble Tube
7500381112 x 124 mm Tube
Catalog number 75003815
Price (SGD)
608.00
EA
Estimated availability date 05-Nov-2025
Add to cart
Capacity:
50 mL Round-bottom Tube
Price (SGD)
608.00
EA
Add to cart
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
Specifications
TypeRotor Tube Adapter
Capacity50 mL Round-bottom Tube
Max. Tube Diameter32 mm
Max. Tube Length126 mm
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link