TX-400 Rotor Adapters
TX-400 Rotor Adapters
Actual product may vary
TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
75003788175, 200, 225, 250 mL Bottle
7500368076 x 5/7 mL Blood Collection Tube
7500368156 x 10/15 mL Blood Collection Tube
7500368215 mL Conical Tube
7500368350 mL Conical Tube
750037001.5/2.0 mL Conical Microtube
7500370330/25 mL DIN Round/Flat-bottom
7500370625 mL Universal
750037933 mL RIA or Round-bottom without Cap
7500379415 mL Blood Collection Tube
Catalog number 75003788
Price (SGD)
1,135.00
EA
Add to cart
Capacity:
175, 200, 225, 250 mL Bottle
Price (SGD)
1,135.00
EA
Add to cart
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)
Specifications
TypeRotor Adapter
Tube Type175, 200, 225, 250 mL Bottle
Capacity175, 200, 225, 250 mL Bottle
Max. Tube Diameter62 mm
Max. Tube Length125 mm
No. per Pack4
Unit SizeEA