1x1 image pixel for data collection
TX-750 Swinging Bucket/BIOShield 1000A Adapters
TX-750 Swinging Bucket/BIOShield 1000A Adapters
Actual product may vary
TX-750 Swinging Bucket/BIOShield 1000A Adapters
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Have Questions?
Catalog NumberCapacity
75003737250 mL Flat-bottom Bottles
Catalog number 75003737
Price (SGD)
1,395.00
EA
Estimated availability date 05-Nov-2025
Add to cart
Capacity:
250 mL Flat-bottom Bottles
Price (SGD)
1,395.00
EA
Add to cart
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
Specifications
TypeSwinging Bucket Adapter
Capacity250 mL Flat-bottom Bottles
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link