Microliter 30 x 2 mL Fixed Angle Rotor for compact and 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Microliter 30 x 2 mL Fixed Angle Rotor for compact and 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Actual product may vary
Thermo Scientific
Microliter 30 x 2 mL Fixed Angle Rotor for compact and 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Perform high-speed separations at 15,200rpm using the Thermo Scientific™ Microliter 30 x 2mL Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
7500365230 x 2 mL
Catalog number 75003652
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
30 x 2 mL
Perform high-speed separations at 15,200rpm using the Thermo Scientific™ Microliter 30 x 2mL Fixed-Angle Rotor.
Compatible with:
Thermo Scientific Sorvall™ Legend™ X1, Sorvall ST 16, Megafuge™ 16, Multifuge™ X1, SL 16 Series
Recommended for:
Supports micro-volume protocols including nucleic acid or protein lysate preparation and PCR reaction setup.
Specifications
DescriptionMicroliter 30 x 2mL Fixed Angle Rotor
TypeFixed Angle Rotor
Capacity30 x 2 mL
Max. RCF25,830 x g
Max. Speed15,200 rpm
Radius (Metric)10 cm
Max. Tube Length50 mm
Max. Tube Diameter11 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge 16, Multifuge X1, SL 16 Series
Angle45°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
BiocontainmentYes
Unit SizeEA