24 x 1.5mL Fixed Angle Rotor
Actual product may vary
Thermo Scientific
24 x 1.5mL Fixed Angle Rotor
Have Questions?
Catalog NumberCapacity
7500333124 x 1.5 mL
Catalog number 75003331
HSN 84219100
Price (SGD)
3,105.00
Each
Capacity:
24 x 1.5 mL
Price (SGD)
3,105.00
Each
Specifications
TypeFixed Angle Rotor
Capacity24 x 1.5 mL
Max. RCF50,377 x g
Max. Speed23,300 rpm
Radius (Metric)0.83 cm
For Use With (Equipment)Thermo Scientific™ Sorvall™ Stratus Centrifuge
Angle45°
Certifications/ComplianceBiocontainment approved
Unit SizeEach