Sorvall™ Tube Adapters for Six-Place Fixed-Angle Rotors
Sorvall™ Tube Adapters for Six-Place Fixed-Angle Rotors
Actual product may vary
Sorvall™ Tube Adapters for Six-Place Fixed-Angle Rotors
Adapt your centrifuge tubes to fit six-place fixed-angle rotor centrifuges with these tube adaptors in various sizes from 1.5 to 50mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Tube Type
75003095HiGhConic 1-place rotor15mL conical bottom tubes
75003091
also known as 75003091FS
HiGhConic 4-place rotor3.5mL round bottom tubes
Catalog number 75003095
Price (SGD)
219.00
EA
Add to cart
For Use With (Equipment):
HiGhConic 1-place rotor
Tube Type:
15mL conical bottom tubes
Price (SGD)
219.00
EA
Add to cart
Adapt your centrifuge tubes to fit Sorvall Legend™ T/RT, Stratos™ and Primo™/Primo R™ centrifuges with these Sorvall™ Tube Adapters for Six-Place Fixed-Angle Rotors. Select from options to accommodate various tube sizes and shapes from 1.5 to 50mL and round or conical bottoms.
  • Legend T/RT and Stratos: 75003334 and 75003046
  • Primo/Primo R: 75007590
  • Accommodate tube sizes from 1.5 to 50mL
Specifications
TypeFixed Angle Rotor
Tube Type15mL conical bottom tubes
Capacity15 mL
For Use With (Equipment)HiGhConic 1-place rotor
Tube Size15 mL
Unit SizeEA