Search Thermo Fisher Scientific
| Catalog Number | DimensionsLxW | Description |
|---|---|---|
| 74018-00 | Mat | |
| FIS14-206-37 | 250 ft. Dispenser Pk. Roll | |
| FIS14-206-38 | Mat | |
| FKS10182361 | Mat | |
| FIS14-206-58 | Table Mat | |
| 74050-00 | Roll | |
| 74218-00 | Mat | |
| FIS14-206-59 | Mat | |
| FKS10325861 | Mat |
Absorb spills and cushion breakable items with Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors. They are long-lasting, waterproof, skid-resistant and chemical-resistant. The cloth-like top layer will not fall apart even when wet.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.