Nalgene™ BioTransport Carrier Handles
Nalgene™ BioTransport Carrier Handles
Actual product may vary
Nalgene™ BioTransport Carrier Handles
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage.
Have Questions?
Catalog NumberDescription
7136-0001Bio Transport Carrier Handle
Catalog number 7136-0001
Price (SGD)
265.00
CS
Add to cart
Description:
Bio Transport Carrier Handle
Price (SGD)
265.00
CS
Add to cart
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage. These handles fit the BioTransport carrier for easy one-handed carrying.
  • Simple to put on and take off
  • Autoclavable stainless steel
  • Includes: Handle only. Biotransport Carrier sold separately, Cat. No. 7135-0001
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Product TypeBioTransport Carrier Handle
DescriptionBio Transport Carrier Handle