Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Actual product may vary
Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Nalgene™ HDPE Pans
Soak labware or form ice baths using these HDPE pans, which are tapered for nesting and chemically resistant to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
7120-002020 L
7120-001010 L
FIS13-359-2510 L
FIS13-359-2620 L
Catalog number 7120-0020
Price (SGD)
496.00
CS
Add to cart
Capacity (Metric):
20 L
Price (SGD)
496.00
CS
Add to cart
Soak labware or form ice baths using Thermo Scientific™ Nalgene™ HDPE Pans. These pans are tapered for nesting and are chemically resistant to most acids, bases and alcohols.
  • Without covers
  • Rugged and durable, tapered for nesting
  • Molded-in handles and rolled edges for easy carrying and handling
  • Resistant to most acids, bases and alcohols for a multitude of lab uses
.
Note:
These pans are NOT autoclavable.
Specifications
Capacity (Metric)20 L
Dimensions (L x W x H)21.375 x 17.125 x 5.125 in. (54 x 43 x 13 cm)
DescriptionRugged, durable pans tapered for nesting
Height (Metric)130.3 mm
Length (Metric)541 mm
MaterialHDPE
TypePan
Width (Metric)434 mm
Length (English)21 in.
No. per Pack1
Height (English)5.13 in.
No. per Case6
ShapeRectangular
Width (English)17.1 in.
AutoclavableNot Autoclavable
Unit SizeCS
Frequently asked questions (FAQs)
Do you offer any trays or pans for secondary containment of reagent bottles?
Yes, 6900 series polypropylene or 7120 series high-density polyethylene trays can be used for this purpose.