RNase AWAY™ Surface Decontaminant
RNase AWAY™ Surface Decontaminant
Actual product may vary
Thermo Scientific
RNase AWAY™ Surface Decontaminant
Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Container Type
7005-114 LBottle
FIS21-236-21250 mLBottle
7002
also known as SP7002A
473 mLSpray Bottle
FIS21-402-178473 mLSpray Bottle
FKS10666421473 mLSpray Bottle
70031 LBottle
FIS14-754-344 LBottle
Catalog number 7005-11
Price (SGD)
261.00
EA
Add to cart
Volume (Metric):
4 L
Container Type:
Bottle
Price (SGD)
261.00
EA
Add to cart
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.
  • Use on pipettes, gel boxes, or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
Specifications
pH11
CompositionSodium Hydroxide with Water
Container TypeBottle
Volume (English)1.06 gal.
Volume (Metric)4 L
Unit SizeEA
Frequently asked questions (FAQs)
Do you have a Technical data sheet (TDS) for RNase AWAY Surface Decontaminant?
We have a safety data sheet (SDS) and COA for RNase AWAY Surface Decontaminant. Visit our Documents and Certificates page to search for and request these documents.