Check Order Status
Filling Tubes for BRAND™ Bottletop Dispensers and Bottletop Burettes
Actual product may vary
Filling Tubes for BRAND™ Bottletop Dispensers and Bottletop Burettes
Replacement filling Tubes for Dispensette™ S / Organic / Trace Analysis Bottletop Dispensers, seripettor™ / seripettor™ pro Bottletop Dispensers, and Titrette™ Bottletop Burettes.
Have Questions?
Catalog Number
Length (Metric)
Sterility
For Use With (Equipment)
70-4538
500 mm
Sterile
Dispensette S (all models), III, Organic, Trace Analysis, and seripettor pro
Catalog number
70-4538
Price (SGD)
227.00
EA
Add to cart
Length (Metric):
500 mm
Sterility:
Sterile
For Use With (Equipment):
Dispensette S (all models), III, Organic, Trace Analysis, and seripettor pro
Price (SGD)
227.00
EA
Add to cart
Available in various lengths depending on the nominal volume of the dispenser
Telescoping filling tubes, made of FEP, adjusts to the correct height without the need for cutting
Standard filling tubes, made of PP, can be manually cut to fit different bottle sizes
Unique design allows liquid aspiration from bottom of bottle while minimizing the intake of precipitates
Telescoping filling tubes are also compatible with Dispensette™ III
Specifications
Color
Clear
Description
Nominal Volume 2, 10, 25 mL
Material
Polypropylene
For Use With (Equipment)
Dispensette S (all models), III, Organic, Trace Analysis, and seripettor pro
Sterility
Sterile
Length (Metric)
500 mm