1x1 image pixel for data collection
Nalgene™ Autoclavable Polypropylene Pans
Nalgene™ Autoclavable Polypropylene Pans
Actual product may vary
Nalgene™ Autoclavable Polypropylene Pans
Nalgene™ Autoclavable Polypropylene Pans
Nalgene™ Autoclavable Polypropylene Pans
Nalgene™ Autoclavable Polypropylene Pans
Repeatedly autoclave instruments and labware with these autoclavable polypropylene pans, featuring heavy-duty construction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
6902-10002 L
FIS13-361-202 L
FIS13-361-152.8 L
6902-30003.1 L
FIS13-361-103.1 L
6902-50005 L
FIS13-361-255 L
Catalog number 6902-1000
Price (SGD)
36.00
EA
Estimated availability date 04-Nov-2025
Add to cart
Capacity (Metric):
2 L
Price (SGD)
36.00
EA
Add to cart
Repeatedly autoclave instruments and labware with Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans, featuring heavy-duty construction, extra strong rims and rounded corners for easy cleaning.
  • Tapered for nesting to save space
  • Rolled edges provide extra stiffness for carrying heavy contents
  • Heavy wall construction will withstand rugged use and repeated autoclaving
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Capacity (Metric)2 L
Dimensions (L x W x H)10.25 x 6.25 x 2.5 in.
DescriptionPolypropylene Autoclavable Pan
Height (Metric)64 mm
Length (Metric)260 mm
MaterialPolypropylene
TypeAutoclavable Pan
Width (Metric)159 mm
Length (English)10 1/4 in.
No. per Pack1
Height (English)2 1/2 in.
Width (English)6 1/4 in.
AutoclavableAutoclavable
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link