Enzyme Freezer Storage Bins
Enzyme Freezer Storage Bins
Actual product may vary
Enzyme Freezer Storage Bins
Enzyme Freezer Storage Bins
Enzyme Freezer Storage Bins
Store and manage inventory effectively with Enzyme Freezer Storage Bins.
Have Questions?
Catalog number 6736
Price (SGD)
139.00
EA
Add to cart
Price (SGD)
139.00
EA
Add to cart
Store and manage inventory effectively with Thermo Scientific™ Enzyme Freezer Storage Bins.
  • High-impact ABS plastic, off-white
  • Interior: 3.5W × 3D × 24in.L (90 × 76 ×610mm)
  • Exterior: 4.4W × 3.04 × 25.25in.L (112 × 77 × 641mm)
  • Labels not included
Specifications
DescriptionABS Plastic Enzyme Freezer Storage Bin
Product TypeKit: Food Analysis (Limited Shelf Life)
Includes1 teacher's guide with MSDS; 10 student worksheets and guides; 2 plastic droppers; 20 slides; 10 test tubes; 1 drop control bottle of Fat Testing solution; 1 drop control bottle each of: Protein Testing solution Type A, Protein Testing solution Type B, Starch Testing solution, Sugar Testing solution, Vitamin C Standardizing solution, and Vitamin C Testing solution
For Use With (Equipment)Storage and inventory management
Class Size10 to 40 Students
Depth (English) Exterior3.04 in.
Depth (Metric) Exterior77 mm
Length (English) Exterior25.25 in.
Length (Metric) Exterior641 mm
Width (English) Exterior112 in.
Width (Metric) Exterior4.4 mm
Unit SizeEA