Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Actual product may vary
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
6696-123.3, 45.8, 51.1 cu.ft. UnitsFull Solid Shelves
6695
also known as JN669X5
29.2 cu. ft. UnitsOpen Wire Shelves
6695-129.2 cu. ft. High-performance UnitsOpen Wire Shelves
669623.3, 45.8, 51.1, 78.8 cu. ft. UnitsOpen Wire Shelves
Catalog number 6696-1
Price (SGD)
505.00
EA
Add to cart
For Use With (Equipment):
23.3, 45.8, 51.1 cu.ft. Units
Type:
Full Solid Shelves
Price (SGD)
505.00
EA
Add to cart
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
  • Warranty varies by country, please contact us for details
Specifications
TypeFull Solid Shelves
Exterior FinishStainless Steel
For Use With (Equipment)23.3, 45.8, 51.1 cu.ft. Units
WarrantyWarranty varies by country, please contact us for details
HoldsGeneral Freezer Items
Unit SizeEA