Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Actual product may vary
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.
Have Questions?
Catalog NumberHeight (Metric)Width (Metric)
6319-004051 mm102 mm
Catalog number 6319-0040
Price (SGD)
413.00
CS
Add to cart
Height (Metric):
51 mm
Width (Metric):
102 mm
Price (SGD)
413.00
CS
Add to cart
Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.
Durable Polyolefin Labels
  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
  • Excellent for labeling microscopic slides— adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack
Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.
.
Specifications
DescriptionPolyolefin Label, 2 x 4
FormatCase
MaterialPolyolefin
Product TypeLabel
Labels per Sheet10
No. per Case500/Cs.
No. per Pack100/Pk.
Width (English)4 in.
Width (Metric)102 mm
Height (English)2 in.
Height (Metric)51 mm
Unit SizeCS