Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Actual product may vary
Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Cryoware Permanent Markers and Pens
Identify samples and storage boxes permanently with these cryoware permanent markers and pens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. per Case
6313-0020
also known as NLG6313-0020
Black24
339993
also known as NN339993
Assorted (Red, Blue, Green, Black)100
FKS11361775Assorted (Red, Blue, Green, Black)100
FIS12-565-237Black10
6313-0010Assorted (Red, Blue, Green, Black)24
FIS13-382-52Assorted (Red, Blue, Green, Black)24
FKS10314551Assorted (Red, Blue, Green, Black)24
FKS10772711Black24
Catalog number 6313-0020
also known as NLG6313-0020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Black
No. per Case:
24
Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.
 Warranty: 90 days
Specifications
Product TypePermanent Marking Pen
Tip StyleExtra Fine
ColorBlack
DescriptionCryoware Permanent Marker
IncludesConsists of four extra-fine-point pens
For Use With (Application)Used for marking cardboard and Polycarbonate
No. per Case24
Quantity4 Pk.
Unit SizeCS